Order Entry
Canada
ContactUsLinkComponent
Dissolved Oxygen Reagent Set, 60 ml sample, Hach
Dissolved Oxygen Reagent Set, 60 ml sample, Hach
Catalog # CA11024-842
Supplier:  Hach
CAS Number:  
undefined
Dissolved Oxygen Reagent Set, 60 ml sample, Hach
Catalog # CA11024-842
Supplier:  Hach
Supplier Number:  2439802
CAS Number:  

Specifications

  • Chemical method:
    Winkler
  • Description:
    Dissolved Oxygen Reagent Set
  • Size:
    60 mL sample
  • Method:
    Winkler
  • Cat. no.:
    CA11024-842
  • Supplier No.:
    2439802

Specifications

About this item

For determination of Dissolved Oxygen by Drop Count Titration

Replacement reagent set for OX-2P Test Kit (1469-00). Contains: Dissolved Oxygen 1, 2, and 3 Reagent Powder Pillows and Sodium Thiosulfate Standard (0.0109 N) for about 100 tests.

Ordering information: Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accommodate these items.