Order Entry
Canada
ContactUsLinkComponent
Chloride Standard Solution, 100 mg/L as Cl- (NIST), 1 L, Hach
Chloride Standard Solution, 100 mg/L as Cl- (NIST), 1 L, Hach
Catalog # CA97021-488
Supplier:  Hach
CAS Number:  
undefined
Chloride Standard Solution, 100 mg/L as Cl- (NIST), 1 L, Hach
Catalog # CA97021-488
Supplier:  Hach
Supplier Number:  2370853
CAS Number:  

Specifications

  • Concentration:
    100 µg/ml
  • Description:
    Chloride
  • Size:
    1 L bottle
  • Cat. no.:
    CA97021-488
  • Supplier No.:
    2370853

Specifications

About this item

For meter calibrations and accuracy checks in chloride determinations

100 mg/L as Cl- (0.00282 N)