Order Entry
Canada
ContactUsLinkComponent
Sodium hydroxide 0.01 N in 2-propanol/water volumetric solution
Flammable
Corrosive
Sodium hydroxide 0.01 N in 2-propanol/water volumetric solution
Catalog # CA11024-734
Supplier:  Hach
CAS Number:  
undefined
Flammable
Corrosive
Sodium hydroxide 0.01 N in 2-propanol/water volumetric solution
Catalog # CA11024-734
Supplier:  Hach
Supplier Number:  67132
CAS Number:  

Specifications

  • Pack type:
    Dropping bottle, marked
  • Size:
    100 ml
  • Cat. no.:
    CA11024-734
  • Supplier No.:
    67132

Specifications

About this item

 :
 :40 g/mol
Density:0.992 g/cm³
UN:2924
ADR:3,III
 :GHS02
 :GHS05
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548