Criterion™ Blotter, Midi Blotting System, Bio-Rad
Supplier: Bio-Rad
Certificates
About this item
The capacity of the Criterion™ Blotter is 2 midi-sized or 4 mini-sized gels
- Transfers in 30 min to 1 hr for most proteins
- Overnight transfer at lower voltages
- Assembly tray for gel soaking, sandwich assembly, and ensuring correct orientation of gels and blots
- Cooling unit is sufficient for absorbing heat during most transfers
- Color-coded, easy-lock cassettes
- Optional cooling coil for precise temperature control
- Overflow chambers keep the laboratory bench dry
Blotters are available with a choice of plate or wire electrodes. Durable plate electrodes consist of a platinum-coated titanium anode and a stainless steel cathode. Wire electrodes are platinum.
The close spacing of the electrodes (4.3 cm) provides a high field strength and allows rapid transfer with one or two cassettes.
Ordering information: All Criterion™ Blotter Systems include a buffer tank with plate or wire electrodes, lid with power cables, two color-coded gel holder cassettes, absorbent filter paper, four foam pads, gel/blot assembly tray, roller, and sealed ice block.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Frequently Bought Together