SpecCheck Gel Secondary Standard Set, Ozone, Mid-Range, Hach
Catalog # CA97026-524
Supplier: Hach
CAS Number:
Specifications
- Concentration:0 - 0.75 mg/L O₃
- Description:SpecCheck Gel Secondary Standard Set, Ozone, Mid-Range
- Method:Indigo
- Instrument:Includes values for all instruments, except DR/3000 and DR/700
- Cat. no.:CA97026-524
- Supplier No.:2708000
Specifications
About this item
For Ozone Indigo method, MR, 0-.75 mg/L as O₃.
- Fast and Convenient
- Stable
Used to confirm consistent instrument response. Set contains four vials filled with gels that simulate the test color at various concentrations.
Ordering information: Set of 4 vials.