ContactUsLinkComponent
Thermometer Probe, Ambient Type J, Sper Scientific
Thermometer Probe, Ambient Type J, Sper Scientific
Catalog # 14003-156
Supplier:  Sper Scientific
undefined
Thermometer Probe, Ambient Type J, Sper Scientific
Catalog # 14003-156
Supplier:  Sper Scientific
Supplier Number:  800097
Specifications
  • Description:
    Ambient Type J Thermometer Probe
  • Environmentally Preferable:
  • Cat. no.:
    14003-156
  • Supplier No.:
    800097
Specifications
About this item
Beaded wire for fast response in air and gas in difficult to access areas
38" cord length.
Ordering information: Six month warranty.