Blanking Reagent for ULR Chlorine, 29 ml, Hach
CA24930-23
: Hach
:
- Chemical method:DPD (N,N-diethyl-p-phenylenediamine)
- Description:Blanking Reagent for ULR Chlorine
- Size:29 mL
- Method:DPD
- Cat. no.:CA24930-23
- Range:2 - 500 µg/L
- Supplier No.:24930-23
- No. of Tests:20
For determination of ultra-low range chlorine by the DPD ULR Method
Range 2 - 500 µg/L, USEPA accepted for reporting wastewater analysis results. *This product has not been evaluated to test for chlorine and chlormaines in medical applications in the United States.
: Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accommodate these items.