F422 HDPE Foam-Lined Polypropylene Caps, Qorpak®
89237-498
: Qorpak
Small Business Enterprise
- Cap Size:38-439
- Cat. no.:89237-498
- Supplier No.:CAP-00379
F422 HDPE Foam Lined Polypropylene Caps are acid resistant
This F422 liner is comprised of an 0.38" LDPE core sandwiched between identical layers of 0.002–0.003" virgin HDPE.