Potassium Hydroxide Standard Solution, 8.00 N, 500 ml, Hach
Catalog # CA282-49
Supplier: Hach
CAS Number:
Specifications
- Description:Potassium Hydroxide Standard Solution, 8.00 N
- Size:500 ml bottle
- Method:EDTA Titration
- Cat. no.:CA282-49
- Supplier No.:282-49
Specifications
About this item
For pH adjustment in calcium determination by EDTA titration
Hach methods 8222 (buret) and 8204 (Digital Titrator).
Ordering information: Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accommodate these items.