Silica Reagent Set, Ultra Low Range, Bulk Solution, Hach
CA11024-850
: Hach
:
- Chemical method:Heteropoly blue
- Description:Silica Reagent Set, Ultra Low Range, Bulk Solution
- Method:8282 - Heteropoly Blue
- Instrument:All except DR/800, PC II
- Cat. no.:CA11024-850
- Range:3 - 1000 µg/L SiO₂
- Supplier No.:2553500
- No. of Tests:100
For determination of ultra-low range Silica by the Heteropoly Blue Method
Hach Method 8282. Range: 3 - 1000 µg/L as SiO2.
: Reagent set includes: Molybdate 3, Citric Acid F Solution and Amino Acid F solution for approximately 100 tests.
Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accomodate these items.
Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accomodate these items.