Order Entry
Canada
ContactUsLinkComponent
Molybdovanadate Reagent, 100 mL, Hach
Molybdovanadate Reagent, 100 mL, Hach
  CA20760-32
 :  Hach
 :  
undefined
Molybdovanadate Reagent, 100 mL, Hach
  CA20760-32
 :  Hach
 :  2076032
 :  

 

  • Chemical method:
    Molybdovanadate
  • Description:
    Molybdovanadate Reagent
  • Size:
    100 mL MDB
  • Method:
    8114 - Molybdovanadate
  • Storage temperature:
    10 to 25 °C
  • Instrument:
    All except DR/850, DR/820, PC II
  • Cat. no.:
    CA20760-32
  • Range:
    0.3 - 45.0 mg/l PO₄
  • Supplier No.:
    20760-32
  • No. of Tests:
    50

 

 

For phosphate (phosphorus) determination by the Molybdovanadate Method: Hach method 8114

Range: up to 45.0 mg/L PO4.

 : Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accomodate these items.