Chlorine (Free and Total) Test Kit, Model CN-70, Hach
Catalog # CA14542-00
Supplier: Hach
CAS Number:
Specifications
- Chemical method:DPD (N,N-diethyl-p-phenylenediamine)
- Description:Chlorine (Free and Total) Test Kit, Model CN-70
- Model:CN-70
- Method:Color disc/DPD
- Case style:D
- Increments:smallest steps - 0.1
- Cat. no.:CA14542-00
- Range:0 - 3.4 mg/l, 0.02 - 0.7 mg/l
- Supplier No.:14542-00
- No. of Tests:100 F, 100 T
Specifications
About this item
Simple, low-cost color disc method is the most accurate visual test kit method.
- Simple, lowcost color disc method is the most accurate visual test kit method
- Achieve fast results with an easytocarry test kit
*This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.
*mg/L unless otherwise noted, ppb = μg/L, ppm = mg/L; gpg = grains per gallon; 1 gpg = 17.1 mg/L or 17.1 ppm.