ContactUsLinkComponent
Dissolved Oxygen AccuVac® Ampules, LR, Hach
Catalog # CA25010-25
Supplier:  Hach
CAS Number:  
undefined
Dissolved Oxygen AccuVac® Ampules, LR, Hach
Catalog # CA25010-25
Supplier:  Hach
Supplier Number:  2501025
CAS Number:  
Specifications
  • Chemical method:
    Indigo carmine
  • Description:
    Dissolved Oxygen AccuVac® Ampules, LR
  • Cat. no.:
    CA25010-25
  • Range:
    6 - 800 μg/l O₂
  • Supplier No.:
    25010-25
  • No. of Tests:
    25
Specifications
About this item
For determination of low range Dissolved Oxygen by the Indigo Carmine Method, using AccuVac® Ampules
  • Fast
  • Easy
  • Accurate
  • Use them anywhere you need results.
Hach Method 8316. Range: 8 - 1000 µg/L O2.
Ordering information: Package of 25 ampules.