Hardness 2 Indicator Solution, 100 ml, Hach
Catalog # CA425-32
Supplier: Hach
CAS Number:
Specifications
- Chemical method:EDTA
- Description:Hardness 2 Indicator Solution
- Size:100 mL Marked Dropping Bottle
- Method:EDTA Titration
- Cat. no.:CA425-32
- Supplier No.:425-32
- No. of Tests:~ 300
Specifications
About this item
ManVer (Calmagite) Hardness Indicator, for hardness and chelant determination by titration.
Ordering information: Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accomodate these items.