Phosphorus (Reactive) TNT Reagent Set, High Range Molybdovanadate, Hach
Catalog # CA11024-870
Supplier: Hach
CAS Number:
Specifications
- Chemical method:Molybdovanadate
- Description:Phosphorus (Reactive) TNT Reagent Set, High Range Molybdovanadate
- Method:8114 - Molybdovanadate
- Instrument:All except DR/850, DR/820, PC II
- Cat. no.:CA11024-870
- Range:1.0 - 100.0 mg/l PO₄
- Supplier No.:2767345
- No. of Tests:25 - 50
Specifications
About this item
For determination of Reactive Phosphorus (ortho-phosphate) by the Molybdovanadate Test 'N Tube™ Method
- Test 'N Tube products provide safe, convenient testing
- Easy ordering
Hach Method 8114. Range: 1.0 - 100.0 mg/L PO4.
Ordering information: Set includes vials and reagents for up to 50 tests (dependent upon the number of reagent blank tests performed).
Hazardous Items:
Items with this mark may be considered hazardous under some shipping conditions.
If necessary, we will change your selected shipping method to accomodate these items.
Hazardous Items:
Items with this mark may be considered hazardous under some shipping conditions.
If necessary, we will change your selected shipping method to accomodate these items.