Order Entry
Canada
ContactUsLinkComponent
Sodium Chloride Standard Solution, 491 mg/L NaCl (1000 µS/cm), Hach
Sodium Chloride Standard Solution, 491 mg/L NaCl (1000 µS/cm), Hach
Catalog # CA14400-42
Supplier:  Hach
CAS Number:  
undefined
Sodium Chloride Standard Solution, 491 mg/L NaCl (1000 µS/cm), Hach
Catalog # CA14400-42
Supplier:  Hach
Supplier Number:  1440042
CAS Number:  

Specifications

  • Concentration:
    491 mg/L NaCl
  • Size:
    100 ml
  • Cat. no.:
    CA14400-42
  • Accuracy:
    ±10 µS/cm
  • Conductivity:
    1000 µS/cm
  • Supplier No.:
    14400-42

Specifications

About this item

For calibrations/accuracy checks on conductivity meters.