NitraVer® 5 Nitrate Reagent Powder Pillows, 5 ml, Hach
Catalog # CA14035-99
Supplier: Hach
CAS Number:
Specifications
- Chemical method:Cadmium reduction
- Description:NitraVer® 5 Nitrate Reagent Powder Pillows
- Size:100 Pillows
- Method:Cadmium Reduction
- Cat. no.:CA14035-99
- Range:up to 50 mg/L as nitrate-N
- Sample Size:5 ml
- Supplier No.:14035-99
Specifications
About this item
For high range nitrate determination by the NitraVer® 5 cadmium reduction method
Range: up to 50 mg/L as nitrate-N.
Caution: Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accommodate these items.