



1-(2-Pyridylazo)-2-naphthol (PAN) solution 0.3% indicator
Catalog # CA21502-32
Supplier: Hach
CAS Number:
Specifications
- Pack type:Bottle
- Size:100 ml
- Cat. no.:CA21502-32
- Supplier No.:21502-32
Specifications
About this item
:
:249.27 g/mol
UN:3082
ADR:9,III
:GHS08
:GHS05
:GHS09
:GHS07
Storage Temperature:Ambient
CAS Number:85-85-8
Formula:C₁₅H₁₁N₃O
MDL Number:MFCD00004071