SPADNS 2 (Arsenic-free) Fluoride Reagent Solution, 500 ml, Hach
CA11016-446
: Hach
:
- Chemical method:SPADNS 2 reagent
- Description:SPADNS 2 (Arsenic-free) Fluoride Reagent Solution
- Size:500 mL bottle
- Method:10225 - SPADNS 2
- Compliance:EPA - Yes
- Instrument:All except DR/820
- Cat. no.:CA11016-446
- Range:0.02 - 2.00 mg/l F-
- Supplier No.:2947549
- No. of Tests:125
For determination of Fluoride by Hach SPADNS 2 Method 10225
-Identical performance to traditional Hach SPADNS
The SPADNS2 reagent formulation removes interference from chlorine without arsenic. Range: 0.02 - 2.00 mg/L F-. EPA compliant.
: Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accommodate these items.