Order Entry
Canada
Orders LinkContactUsLinkComponent
Starch indicator in solution redox indicator

Starch indicator in solution redox indicator

MDB, 100 ml
Catalog # CA349-32
Supplier:   Hach
Supplier #:  34932
Starch indicator in solution redox indicator
Catalog # CA349-32
Supplier:  Hach
CAS Number:  9005-25-8
Starch indicator in solution redox indicator
Starch indicator in solution redox indicator

MDB, 100 ml

CAS Number:  9005-25-8
Price
$40.81
Log in for availability
Each (100ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Oxidation-Reduction Indicator for iodometric titrations.

  • CAS Number:9005-25-8
  • Formula:(C₆H₁₀O₅)n
  • MDL Number:MFCD00082026
  • Molecular Weight:162.14 g/mol
  • Storage Temperature:Ambient
Specifications

Specification Test Results

  • Application:
    Remove Chlorine Interference
  • Description :
    Remove Interfering substance
  • Formula:
    Titration Indicator
  • Number of tests:
    Varies
  • Package Type:
    Bottle
  • Platform :
    Solution
  • Quantity:
    100 mL

Specifications

  • Pack type:
    MDB
  • Size:
    100 ml

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.