ContactUsLinkComponent
Silica, Low Range, Reagent Set, Hach
Silica, Low Range, Reagent Set, Hach
  CA2459300
 :  Hach
 :  
Silica, Low Range, Reagent Set, Hach
  CA2459300
 :  Hach
 :  2459300
 :  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Chemical method:
    Heteropoly blue
  • Description:
    Silica, Low Range, Reagent Set
  • Method:
    8186 - Heteropoly Blue
  • Instrument:
    All except DR/3000, DR/2000, DR/820, PC II
  • Cat. no.:
    CA2459300
  • Range:
    0.010 - 1.600 mg/l SiO₂
  • Supplier No.:
    2459300
  • No. of Tests:
    100
For determination of Low Range Silica by the Heteropoly Blue method
Hach Method 8186. Range: 0.01 - 1.60 mg/L SiO2. Sample Size: 10 mL. Reagent Set includes Amino Acid F Reagent Powder Pillows, Citric Acid Powder Pillows and Molybdate 3 Reagent Solution for about 100 tests.
 : Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accomodate these items.