About this item
Process a range of general-purpose applications including cell culture, microplate, bioproduction, blood separation, and microbiology with the Multifuge™ X3 Centrifuge Series
To facilitate simple operation, each unit features intuitive control panel and digital display. Detailed operation information, including display of set and actual parameters, maximize run performance and temperature accuracy for improved sample protection. The user can easily adjust speed, time, and temperature, even during the run, with a convenient keyboard for direct input. Innovative lid locking technology enables one finger downward motion for simple closing and locking of centrifuge lid, even on tall benches (with optional automatic lid opening). A double temperature management system displays the air temperature in the chamber as well as the sample temperature, ensuring accurate control of the sample temperature during the run.
Simple Operation:
- System offers intuitive control panel and digital display.
- Detailed operation information, including display of set and actual parameters, maximize run performance and temperature accuracy for improved sample protection.
- User can easily adjust speed, time, and temperature, even during the run, with a convenient keyboard for direct input.
- Innovative lid locking technology enables one finger downward motion for easiest closing and locking of centrifuge lid, even on tall benches with optional automatic lid opening.
- Double temperature management system displays the air temperature in the chamber as well as the sample temperature, ensuring accurate control of the sample temperature during the run.
Thermo Scientific Auto-Lock™ III Rotor System:
- Secure locking system allows easy push-button installation and exchange of rotors.
- System saves time in switches between applications.
- User has easy access to the chamber, enabling quick cleaning, a healthier working environment, and longer unit life.
Featuring industry-leading throughput and energy efficient operation, this series can process up to 40% more conical tubes in swing-out rotor configuration and 100% more in fixed-angle configuration. A broad range of rotors and adapters, including lightweight Fiberlite™ carbon fiber rotors, handle diverse laboratory applications.
Specifications
- Catalog No:
- 10324-264
- 89402-980
- Acceleration/braking profiles:
- Control:
- Functions:
- Memory:
- Display:
- Temperature setting range:
- Depth:
- Width: