Order Entry
Canada
ContactUsLinkComponent
Chloride 2 Indicator Powder Pillows, 100 ml, Hach
Chloride 2 Indicator Powder Pillows, 100 ml, Hach
Catalog # CA1057-66
Supplier:  Hach
CAS Number:  
Chloride 2 Indicator Powder Pillows, 100 ml, Hach
Catalog # CA1057-66
Supplier:  Hach
Supplier Number:  105766
CAS Number:  

Specifications

  • Chemical method:
    Silver nitrate
  • Description:
    Chloride 2 Indicator Powder Pillows
  • Size:
    100 mL samlple
  • Method:
    8207; 8225 - Silver Nitrate Titration
  • Cat. no.:
    CA1057-66
  • Supplier No.:
    1057-66
  • No. of Tests:
    50

Specifications

About this item

Indicator used in the determination of chloride by argentometric titration with silver nitrate.

Ordering information: Pack of 50 Powder Pillows.

Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accomodate these items.