Dissolved Oxygen Test Kit, Model OX-2P, Hach
Catalog # CA1469-00
Supplier: Hach
CAS Number:
Specifications
- Chemical method:Winkler
- Description:Dissolved Oxygen Test Kit, Model OX-2P
- Model:OX-2P
- Method:Drop count titration/ Modified Winkler
- Case style:D
- Cat. no.:CA1469-00
- Range:1 - 20 mg/l O₂, 0.2 - 4 mg/l O₂
- Supplier No.:1469-00
- No. of Tests:100
Specifications
About this item
Low dissolved oxygen levels in surface water systems usually indicate serious pollution
Utilizes drop count titration - a simple, economical titration method.
Adequate amounts are crucial for fish life, but conversely, DO must be excluded from boiler feed water to prevent corrosion. This test kit utilizes drop count titration and contains 100 tests.
Ordering information: Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accomodate these items.