Order Entry
Canada
ContactUsLinkComponent
EDTA standard solution 0.0200N
EDTA standard solution 0.0200N
  CA205-53
 :  Hach
 :  
undefined
EDTA standard solution 0.0200N
  CA205-53
 :  Hach
 :  20553
 :  

 

  • Pack type:
    Plastic bottle
  • Size:
    1 L
  • Cat. no.:
    CA205-53
  • Supplier No.:
    205-53

 

 

 :
 :380.17 g/mol
 :64-02-8
 :C₁₀H₁₂N₂Na₄O₈
 :MFCD00150027