ChromaVer® 3 Chromium Reagent Powder Pillows, 5 or 10 ml, Hach
Catalog # CA12710-99
Supplier: Hach
CAS Number:
Specifications
- Chemical method:Diphenylcarbazide
- Description:ChromaVer® 3 Chromium Reagent Powder Pillows, 5 or 10 mL
- Compliance:EPA - Yes
- Cat. no.:CA12710-99
- Range:0.010 - 0.700 mg/L Cr6+
- Supplier No.:12710-99
- No. of Tests:100
- For Unit:All except DR/3000, DR/2000, DR/850, DR/820
Specifications
About this item
For determination of hexavalent chromium by the Diphenylcarbohydrazide method
USEPA accepted for reporting for wastewater analyses using Hach Method 8023, Diphenylcarbohydrazide Method. Photometric Sample Size: 10 mL. Photometric Range: up to 0.70 mg/L. Kit Sample Size: 5 mL. Kit Range: Up to 1.50.
Ordering information: Pack of 100 powder pillows.