Order Entry
Canada
ContactUsLinkComponent
Molybdenum Reagent Set, HR, 10 ml, Hach
Molybdenum Reagent Set, HR, 10 ml, Hach
Catalog # CA26041-00
Supplier:  Hach
CAS Number:  
undefined
Molybdenum Reagent Set, HR, 10 ml, Hach
Catalog # CA26041-00
Supplier:  Hach
Supplier Number:  2604100
CAS Number:  

Specifications

  • Chemical method:
    Mercaptoacetic acid
  • Description:
    Molybdenum Reagent Set, HR
  • Size:
    10 mL Sample
  • Method:
    8036 - Mercaptoacetic Acid
  • Instrument:
    All except DR/3000, DR/2000, DR/850, DR/820, PC II
  • Cat. no.:
    CA26041-00
  • Range:
    0.3 - 40.0 mg/l Mo
  • Supplier No.:
    2604100
  • No. of Tests:
    100

Specifications

About this item

For determination of Molybdate Molybdenum by the Mercaptoacetic Acid Method

Hach method 8036. Range: 0.3 - 40.0 mg/L Mo.

Ordering information: Reagent set includes MolyVer® 1, 2 and 3 Molybdenum Reagent Powder Pillows for 100 tests.