Chlorine Reagent Set (Total), Ultra Low Range DPD, Hach
Catalog # CA11024-852
Supplier: Hach
CAS Number:
Specifications
- Chemical method:DPD (N,N-diethyl-p-phenylenediamine)
- Description:Chlorine Reagent Set (Total), Ultra Low Range DPD
- Method:8370 - DPD
- Compliance:EPA - Yes
- Instrument:All except DR/800, PC II
- Cat. no.:CA11024-852
- Range:2 - 500 μg/l Cl₂
- Supplier No.:2563000
- No. of Tests:20
Specifications
About this item
For determination of ultralow Total Chlorine concentrations by the DPD Method
*This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.
Hach Method 8370. USEPA accepted for drinking water and wastewater analysis. Range: 2 - 500 μg/L Cl2. Reagents are ampuled for stability.
Ordering information: Reagent set includes ULR Chlorine Buffer Solution (1.5-mL ampules), DPD Indicator Solution for ULR Chlorine (1.5-mL ampules) and Blanking Reagent for 20 tests.
Hazardous Items:
Items with this mark may be considered hazardous under some shipping conditions.
If necessary, we will change your selected shipping method to accomodate these items.
Hazardous Items:
Items with this mark may be considered hazardous under some shipping conditions.
If necessary, we will change your selected shipping method to accomodate these items.