Citric Acid F Reagent Solution, 100 ml, Hach
CA22542-32
: Hach
:
- Chemical method:Heteropoly blue
- Description:Citric Acid F Reagent Solution
- Size:100 mL bottle
- Method:8282 - Heteropoly Blue
- Cat. no.:CA22542-32
- Range:3-1000 μg/L SiO₂
- Supplier No.:22542-32
- No. of Tests:100
For determination of ultra-low range silica by the Heteropoly Blue method (Hach Method 8282)
Range: 3-1000 μg/L SiO2.
: Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accomodate these items.