Order Entry
Canada
ContactUsLinkComponent
 
Diphenylcarbazone Reagent Powder Pillows, 100 ml sample, Hach
  CA836-99
 :  Hach
 :  
undefined
Diphenylcarbazone Reagent Powder Pillows, 100 ml sample, Hach
  CA836-99
 :  Hach
 :  83699
 :  

 

  • Chemical method:
    Mercury nitrate
  • Description:
    Diphenylcarbazone Reagent Powder Pillows
  • Size:
    100 mL Sample
  • Method:
    8206 - Mercuric Nitrate Titration
  • Cat. no.:
    CA836-99
  • Supplier No.:
    836-99
  • No. of Tests:
    100

 

 

Indicator-buffer reagent for chloride determination by Hach mercuric nitrate titration methods

Hach method 8206. Powder pillows for 100-mL sample.

 : Pack of 100 powder pillows.