Distillation Head, Reflux, Ace Glass
Catalog #: 89074-384
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- ST Joint:14/20
- Description:Distillation Head (Complete)
- Hose connection int.Ø:5/16" tubing
- Cat. no.:89074-384
- Thermometer ST Joint Size:10/30
- Supplier No.:9358-03
Specifications
About this item
This distillation head has a cold finger type condenser and 0-3mm PTFE stopcock plug with PTFE ring seals for reflux control
The seals prevent o-ring exposure to corrosive liquids, and they are backed up by a FETFE o-ring. Fabricated from borosilicate glass.
Ordering information: Complete item comes with 89074-438 distillation head and 89074-306 cold finger condenser. Individual components may be purchased separately.
Customers Who Bought This Also Bought