Order Entry
Canada
ContactUsLinkComponent
Molybdenum Reagent Set, LR, 20 ml, Hach
Molybdenum Reagent Set, LR, 20 ml, Hach
  CA11014-442
 :  Hach
 :  
undefined
Molybdenum Reagent Set, LR, 20 ml, Hach
  CA11014-442
 :  Hach
 :  2449400
 :  

 

  • Chemical method:
    Ternary complex
  • Description:
    Molybdenum Reagent Set, LR
  • Size:
    20 mL Sample
  • Method:
    8169 - Ternary Complex
  • Instrument:
    All except DR/3000, DR/2000, DR/820
  • Cat. no.:
    CA11014-442
  • Range:
    0.02 - 3.00 mg/l Mo
  • Supplier No.:
    2449400
  • No. of Tests:
    100

 

 

For determination of low range Molybdate Molybdenum by the Ternary Complex method

Hach Method 8169. Range: 0.02 - 3.00 mg/L Mo.

 : Reagent Set includes: Molybdenum 1 Reagent (LR) Molybdate Powder Pillows and Molybdenum 2 Reagent Solution for approximately 100 tests.