ContactUsLinkComponent
Phosphorus (Reactive) TNT Reagent Set, Low Range, Hach
Phosphorus (Reactive) TNT Reagent Set, Low Range, Hach
Catalog # CA97026-534
Supplier:  Hach
CAS Number:  
undefined
Phosphorus (Reactive) TNT Reagent Set, Low Range, Hach
Catalog # CA97026-534
Supplier:  Hach
Supplier Number:  2742545
CAS Number:  
Specifications
  • Chemical method:
    Ascorbic acid
  • Description:
    Phosphorus (Reactive) TNT Reagent Set, Low Range
  • Method:
    8048 - Ascorbic Acid
  • Instrument:
    All except DR/820, PC II
  • Cat. no.:
    CA97026-534
  • Range:
    0.06 - 5.00 mg/l PO₄
  • Supplier No.:
    2742545
  • No. of Tests:
    25 - 50
Specifications
About this item
For determination of Reactive Phosphorus (ortho-phosphate) by the PhosVer® 3 Ascorbic Acid Test 'N Tube™ method.
  • Test 'N Tube products provide safe, convenient testing
  • Easy ordering
Hach Method 8048. Range: 0.06 - 5.00 mg/L PO4. Set includes: PhosVer® 3 Phosphate Reagent Powder Pillows and Reactive Phosphorus Test 'N Tube Dilution Vials for up to 50 tests.