Order Entry
Canada
Orders LinkContactUsLinkComponent
Timer/Control

Timer/Control

Repeat cycle, electronic timer/control, 120 V, 114.3×158.7×63½ mm (4½×6¼×2½"), 1 lbs. (10 oz.)
Catalog # 89084-418
Supplier:   Ace Glass
Supplier #:  6671-14
Timer/Control
Catalog # 89084-418
Supplier:  Ace Glass
Timer/Control
Timer/Control

Repeat cycle, electronic timer/control, 120 V, 114.3×158.7×63½ mm (4½×6¼×2½"), 1 lbs. (10 oz.)

Price
$5,609.89
New Customer Promotional Price
Log in for availability
Each
Product Details & Documents

This repeat cycle timer control, bench top or rack mountable (has a clamp for use with up to a ¾" rod), is used to regulate the reflux ratio on distillation heads.

  • Maximum ON/OFF time setting is 0.1 to 6000 seconds
  • Solid state relay and micro-processor-based, timer control with LCD display
  • Red output indicator LED

Features include: front toggle main power switch; fast acting, solid state 10 A fuse; rear power outlet; run/standby-set switch on front; four-digit adjust buttons. Light gray case with black front panel.

Delivery information: Comes complete with 6' grounded neoprene line cord with NEMA plug and operating instructions on the case. Conditional two-year warranty. Contact your VWR representative for details. Available in 120 or 220 V versions (both are 10 A maximum, 50/60 Hz).

Specifications
  • Description:
    Repeat cycle, electronic timer/control, 120 V
  • W×H×D:
    114.3×158.7×63½ mm (4½×6¼×2½")
  • Weight:
    1 lbs. (10 oz.)
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.