Orders LinkContactUsLinkComponent
Radioactive Decay Cubes Lab Activity
Radioactive Decay Cubes Lab Activity
Radioactive Decay Cubes Lab Activity, 10
Catalog # 470190-688
Supplier:   Avantor
Radioactive Decay Cubes Lab Activity
Catalog # 470190-688
Supplier:  Avantor
Radioactive Decay Cubes Lab Activity
Radioactive Decay Cubes Lab Activity
Radioactive Decay Cubes Lab Activity, 10
Price
$173.00
Log in for availability
Each Pallet of 100
Product Details & Documents
Radioactive Decay Cubes Lab Activity
  • Determine the half-life of two different "elements"
  • Graphically represent the observed decay
Radioisotopes have been found to be useful in the fields of geology, medical diagnosis and treatment, metallurgy, agriculture, and scientific research. In this lab activity, students will use the framework concepts of Matter and Its Interactions (PS1) to explore the concepts of radioactive decay and half-life. Students will also be able to investigate what happens when a parent nuclei decays into daughter nuclei over time.
This kit has been aligned with all published National Standards. Pre- and Post-laboratory assessments and vocabulary words all target specific Science and Engineering Practices and common core standards. The teacher guide also contains specific "Did You Know" concepts that link the real world with the scientific standards explored. A student copy master is also included. This lab activity can be successfully performed repeatedly with the materials provided and includes enough materials for 10 setups. Time requirements: This lab activity requires approximately one 45-minute class period to complete.
Ordering information: Kit includes 500 Cubes with 2 dots, blue; 500 Cubes with 3 dots, orange; 10 shaker cups with lids; 1 Graph paper pad, 100 sheets; and Teacher's Guide and Student Guide.
Specifications
  • Description:
    Radioactive Decay Cubes Lab Activity
  • No. of lab groups:
    10