If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Chemical method:Ascorbic acid
- Description:PhosVer® 3 Phosphate Reagent Powder Pillows
- Size:5 mL Sample
- Method:Ascorbic Acid
- Cat. no.:CA2209-99
- Supplier No.:2209-99
- No. of Tests:100
For phosphate (phosphorus) determination by the PhosVer 3 ascorbic acid method
For use with visual test kits.