ContactUsLinkComponent
Lauryl Tryptose Broth Tubes, Hach
Lauryl Tryptose Broth Tubes, Hach
Catalog # CA21623-15
Supplier:  Hach
CAS Number:  
undefined
Lauryl Tryptose Broth Tubes, Hach
Catalog # CA21623-15
Supplier:  Hach
Supplier Number:  2162315
CAS Number:  
Specifications
  • Description:
    Lauryl Tryptose Broth Tubes
  • Size:
    pk/15
  • Cat. no.:
    CA21623-15
  • Supplier No.:
    21623-15
Specifications
About this item
Lauryl Tryptose Broth Tubes, single strength, pk/15
Save time and labor costs with ready to use MPN tubes, with minimal training requirements.
Selectivity: Total Coliform Bacteria (presumptive).