Phenol red solution, Spectrophotometric Grade pH-indicator
Catalog # CA97026-490
Supplier: Hach
CAS Number:
Specifications
- Size:50 ml
- Cat. no.:CA97026-490
- Supplier No.:2657512
Specifications
Specification Test Results
- Method Name:Colorimetric Phenol Red
- Number of tests:50 mL
- Parameter:pH
- Platform :Solution
- Quantity:50
- Range:6.5 - 8.5
Specification Test Results
About this item
:
:354.38 g/mol
Storage Temperature:Ambient
CAS Number:143-74-8
Formula:C₁₉H₁₄O₅S
MDL Number:MFCD00003552