Orders LinkContactUsLinkComponent
Microscope Pen, Sper Scientific
Microscope Pen, Sper Scientific
Microscope Pen
Catalog # 97035-716
Microscope Pen, Sper Scientific
Catalog # 97035-716
Supplier:  
Microscope Pen, Sper Scientific
Microscope Pen, Sper Scientific
Microscope Pen
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
POCKET MEASURING MICROSCOPE 50 Catalog #100491-450
Product 100491-450 is similar to 97035-716 but manufacturer and specifications may vary.
Product Details & Documents
Useful for on the spot inspections in printing, textile, art, jewelry, microelectronics, and other industries
  • Also serves as an inexpensive and simple to use microscope for science education and other labs
  • Equipped with powerful 50x optics
  • Weighing only 20g (0.7oz.), the pen clips easily to a shirt pocket and can be carried anywhere
135x10 mm (53/8 x 3/8")
Ordering information: The microscope pen is supplied with a 5-year warranty.
Specifications
  • Description:
    Microscope Pen