Helicoverpa assulta; Heliothis zea Pheromone Biosynthesis Activating Neuropeptide
H-5774.0500BA
:
- CAS No.:122071-54-9
- Species:Helicoverpa assultaHeliothis zea
- Size:0.5 mg
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD00147091
- Protein synonyms:HeA-PBAN|HeZ-PBAN
- Protein/peptide name:Pheromone Biosynthesis Activating Neuropeptide
- Molecular weight:3901.31
- Sequence:H-Leu-Ser-Asp-Asp-Met-Pro-Ala-Thr-Pro-Ala-Asp-Gln-Glu-Met-Tyr-Arg-Gln-Asp-Pro-Glu-Gln-Ile-Asp-Ser-Arg-Thr-Lys-Tyr-Phe-Ser-Pro-Arg-Leu-NH₂
- Molecular formula:C₁₆₇H₂₅₉N₄₇O₅₇S₂
- Cat. no.:H-5774.0500BA
- Supplier No.:H-5774.0500
A pheromone biosynthesis activating neuropeptide hormone which in the picomolar range induced sex pheromone synthesis in various moth species.