H-Arg-Arg-Arg-Arg-Arg-Arg-OH Trifluoroacetate, Bachem
Supplier: Bachem Americas
Certificates
About this item
Further oligoarginines: diarginine (G-1465), triarginine (H-1790), and tetraarginine (H-4464 and H-7354). See also H-6284, FLAG epitope peptide.
Six arginines is the minimum number of residues required for obtaining a polycationic peptide which can effectively penetrate cells.
Specifications
- CAS No.:96337-25-6
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD18633393
- Protein synonyms:H(-Arg)6-OH|Hexaarginine
- Protein/peptide name:H-Arg-Arg-Arg-Arg-Arg-Arg-OH Trifluoroacetate
- Molecular weight:955.14
- Sequence:H-Arg-Arg-Arg-Arg-Arg-Arg-OH
- Molecular formula:C₃₆H₇₄N₂₄O₇
Specifications
Frequently Bought Together