Order Entry
Canada
ContactUsLinkComponent
 
Bovine;Guinea pig;Human;Mouse;Rat (Ser⁸)-GLP-1 (7-36) Amide Trifluoroacetate
 
Bovine;Guinea pig;Human;Mouse;Rat (Ser⁸)-GLP-1 (7-36) Amide Trifluoroacetate

 

  • CAS No.:
    215777-46-1
  • Species:
    Human
    Bovine
    Guinea pig
    Rat
    Mouse
  • Storage conditions:
    –20 ± 5 °C
  • MDL No.:
    MFCD02266190
  • Protein synonyms:
    (Ser8)-Glucagon-like Peptide 1 (7-36) amide|(Ser99)-Preproglucagon (98-127) amide|(Ser79)-Proglucagon (78-107) amide
  • Protein/peptide name:
    (Ser⁸)-GLP-1 (7-36) amide Trifluoroacetate
  • Molecular weight:
    3313.68
  • Sequence:
    H-His-Ser-Glu-Gly-Thr-Phe-Thr-Ser-Asp-Val-Ser-Ser-Tyr-Leu-Glu-Gly-Gln-Ala-Ala-Lys-Glu-Phe-Ile-Ala-Trp-Leu-Val-Lys-Gly-Arg-NH₂
  • Molecular formula:
    C₁₄₉H₂₂₆N₄₀O₄₆

 

Frequently Bought Together