Erythropoietin Mimetic Peptide Sequence 20 Trifluoroacetate
Catalog # H-4344.0001BA
Supplier: Avantor
CAS Number:
Specifications
- CAS No.:203397-62-0
- Source:
- Species:
- Size:1 mg
- Environmentally Preferable:
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD18633330
- Protein synonyms:EMP20
- Protein/peptide name:Erythropoietin Mimetic Peptide Sequence 20 Trifluoroacetate
- Molecular weight:1538.81
- Sequence:H-Tyr-Ser-Cys-His-Phe-Gly-Pro-Leu-Thr-Trp-Val-Cys-Lys-OH
- Molecular formula:C₇₂H₉₉N₁₇O₁₇S₂
- Cat. no.:H-4344.0001BA
- Supplier No.:H-4344.0001
Specifications
About this item
Peptide mimetic of erythropoietin (EPO). EMP-20 has been described to be an excellent starting point for the design of compounds with erythropoietin (EPO) mimetic activity and can serve as a minimal model for an EPO receptor antagonist.