(Des-Gly¹⁰,D-Ser(tBu)⁶,Pro-NHEt⁹)-LHRH Acetate
Supplier: Bachem Americas
Certificates
About this item
LHRH is also called Gonadotropin-Releasing Hormone (GnRH) or Luteinizing Hormone-Releasing Factor (LRF).
Buserelin is a longer acting synthetic peptide analog of the naturally occurring gonadotropin-releasing hormone (GnRH/LHRH). Substitution of glycine in position 6 by D-serine and that of glycinamide in position 10 by ethylamide, leads to a nonapeptide with a significantly enhanced LHRH effect. The effects of buserelin on follicle-stimulating hormone (FSH) and luteinizing hormone (LH) release are approximately 20 to 170 times higher than those of natural LHRH. CAS Number (buserelin acetate): 68630-75-1.
Specifications
- CAS No.:57982-77-1
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD01716229
- Protein synonyms:Buserelin
- Protein/peptide name:(Des-Gly¹⁰,D-Ser(tBu)⁶,Pro-NHEt⁹)-LHRH Acetate
- Molecular weight:1239.44
- Sequence:Pyr-His-Trp-Ser-Tyr-D-Ser(tBu)-Leu-Arg-Pro-NHEt
- Molecular formula:C₆₀H₈₆N₁₆O₁₃
Specifications
Frequently Bought Together