Order Entry
Canada
Orders LinkContactUsLinkComponent
H-Val-Lys-Lys-Arg-OH Acetate

H-Val-Lys-Lys-Arg-OH Acetate

5 mg
Catalog # H-1218.0005BA
H-Val-Lys-Lys-Arg-OH Acetate
Catalog # H-1218.0005BA
Supplier:  
CAS Number:  
H-Val-Lys-Lys-Arg-OH Acetate

5 mg

CAS Number:  
Discontinued
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Specifications

Specifications

  • Size:
    5 mg
  • Cat. No.:
    H-1218.0005BA
  • Molecular formula:
    C₂₃H₄₇N₉O₅
  • Protein/peptide name:
    H-Val-Lys-Lys-Arg-OH Acetate
  • Storage conditions:
    –20 ± 5 °C
  • MDL No.:
    MFCD00238276
  • Sequence:
    H-Val-Lys-Lys-Arg-OH
  • Molecular weight:
    529.68
  • CAS No.:
    286471-39-4

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.