Order Entry
Canada
ContactUsLinkComponent
Recombinant HIV-1 Protease (from E. coli)
Recombinant HIV-1 Protease (from E. coli)
  H-9040.0100BA
 :  
Recombinant HIV-1 Protease (from E. coli)
  H-9040.0100BA
 :  4030739.0100
 :  

 

  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Size:
    0.1 mg
  • Storage conditions:
    <−70 °C
  • Enzyme name:
    HIV-1 Protease
  • Cat. no.:
    H-9040.0100BA
  • Supplier No.:
    H-9040.0100