Order Entry
Canada
Orders LinkContactUsLinkComponent
Substance P (1-7)

Substance P (1-7)

Supplier:   Bachem Americas
Substance P (1-7)
Supplier:  Bachem Americas
Certificates

2 Options Available Below

  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

This amino-terminal fragment of substance P exerts depressor and bradycardic effects when applied to the nucleus tractus solitarius. It was also shown that this fragment, contrary to substance P, may reduce signs of opioid withdrawal in mice.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • H-1582.0005BA
  • H-1582.0025BA
  • Protein/peptide name:
  • Substance P (1-7)
  • Substance P (1-7)
  • CAS No.:
  • 68060-49-1
  • 68060-49-1
  • MDL No.:
  • MFCD00076789
  • MFCD00076789
  • Sequence:
  • Arg-Pro-Lys-Pro-Gln-Gln-Phe
  • Arg-Pro-Lys-Pro-Gln-Gln-Phe
  • Molecular weight:
  • 900.05
  • 900.05
  • Molecular formula:
  • C₄₁H₆₅N₁₃O₁₀
  • C₄₁H₆₅N₁₃O₁₀
  • Storage conditions:
  • –20 ± 5 °C
  • –20 ± 5 °C

Specifications

Product Family Options

Product Information

  • Size
    Availability
    Price
  • 5 mg
    Log in for availability
    Log in for availability
    Each (5mg)
    Each (5mg) - $320.65
    $320.65
    Each (5mg)
       $320.65
    Minimum order quantity is 1

    Supplier Number: 4019809.0005
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    H-1582.0005BA
    Protein/peptide name
    Substance P (1-7)
    CAS No.
    68060-49-1
    MDL No.
    MFCD00076789
    Sequence
    Arg-Pro-Lys-Pro-Gln-Gln-Phe
    Molecular weight
    900.05
    Molecular formula
    C₄₁H₆₅N₁₃O₁₀
    Storage conditions
    –20 ± 5 °C
  • 25 mg
    Log in for availability
    Log in for availability
    Each (25mg)
    Each (25mg) - $1,282.56
    $1,282.56
    Each (25mg)
       $1,282.56
    Minimum order quantity is 1

    Supplier Number: 4019809.0025
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    H-1582.0025BA
    Protein/peptide name
    Substance P (1-7)
    CAS No.
    68060-49-1
    MDL No.
    MFCD00076789
    Sequence
    Arg-Pro-Lys-Pro-Gln-Gln-Phe
    Molecular weight
    900.05
    Molecular formula
    C₄₁H₆₅N₁₃O₁₀
    Storage conditions
    –20 ± 5 °C