Order Entry
Canada
Orders LinkContactUsLinkComponent
Cyclo(-Phe-Phe)

Cyclo(-Phe-Phe)

Supplier:   Bachem Americas
Cyclo(-Phe-Phe)
Supplier:  Bachem Americas
Certificates

2 Options Available Below

  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Also known as dioxopiperazines, piperazine-2,5-diones or DKPs. Diketopiperazines may occur as by-products during peptide synthesis or during the degradation of peptides. These cyclic dipeptides have been detected as taste-modulating compounds in food, they often show biological activity. DKPs are valuable chiral synthons, employed e.g. in Schöllkopf's versatile bislactim ether approach. They also have found use as catalysts for enantioselective synthesis, e.g. in the asymmetric Strecker reaction. See also the TRH metabolite cyclo(-His-Pro), G-1745, and cyclo(-Asp-Phe), G-1695, the major degradation product of aspartame.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • G-1765.0001BA
  • G-1765.0005BA
  • Protein/peptide name:
  • Cyclo(-Phe-Phe)
  • Cyclo(-Phe-Phe)
  • CAS No.:
  • 2862-51-3
  • 2862-51-3
  • MDL No.:
  • MFCD00190985
  • MFCD00190985
  • Sequence:
  • Cyclo(-Phe-Phe)
  • Cyclo(-Phe-Phe)
  • Molecular weight:
  • 294.35
  • 294.35
  • Molecular formula:
  • C₁₈H₁₈N₂O₂
  • C₁₈H₁₈N₂O₂
  • Storage conditions:
  • –20 ± 5 °C
  • –20 ± 5 °C

Specifications

Product Family Options

Product Information

  • Size
    Availability
    Price
  • 1 g
    Log in for availability
    Log in for availability
    Each (1g)
    Each (1g) - $908.47
    $908.47
    Each (1g)
       $908.47
    Minimum order quantity is 1

    Supplier Number: 4002860.0001
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    G-1765.0001BA
    Protein/peptide name
    Cyclo(-Phe-Phe)
    CAS No.
    2862-51-3
    MDL No.
    MFCD00190985
    Sequence
    Cyclo(-Phe-Phe)
    Molecular weight
    294.35
    Molecular formula
    C₁₈H₁₈N₂O₂
    Storage conditions
    –20 ± 5 °C
  • 5 g
    Log in for availability
    Log in for availability
    Each (5g)
    Each (5g) - $3,636.73
    $3,636.73
    Each (5g)
       $3,636.73
    Minimum order quantity is 1

    Supplier Number: 4002860.0005
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    G-1765.0005BA
    Protein/peptide name
    Cyclo(-Phe-Phe)
    CAS No.
    2862-51-3
    MDL No.
    MFCD00190985
    Sequence
    Cyclo(-Phe-Phe)
    Molecular weight
    294.35
    Molecular formula
    C₁₈H₁₈N₂O₂
    Storage conditions
    –20 ± 5 °C
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.