Distillation Receiver, Centrifugal Type, Ace Glass
89074-454
Product available on GSA Advantage®
Small Business Enterprise
- Capacity:15 mL (total)
- Description:Distillation Receiver
- Cat. no.:89074-454
- Top ST Joint:[ST] 14/20, outer
- Subdivisions:0.1 mL
- Supplier No.:9397-06
This distillation receiver is centrifugal type, calibrated to contain
Fabricated from borosilicate glass.