Order Entry
Canada
ContactUsLinkComponent
Citric Acid Reagent Powder Pillows for Silica, 10 ml, Hach
Citric Acid Reagent Powder Pillows for Silica, 10 ml, Hach
Catalog # CA21062-69
Supplier:  Hach
CAS Number:  
undefined
Citric Acid Reagent Powder Pillows for Silica, 10 ml, Hach
Catalog # CA21062-69
Supplier:  Hach
Supplier Number:  2106269
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Citric Acid Reagent Powder Pillows for Silica
  • Size:
    10 mL
  • Method:
    8185, 8186
  • Cat. no.:
    CA21062-69
  • Supplier No.:
    21062-69
  • No. of Tests:
    50 - 100

Specifications

About this item

For determination of high range silica by the silicomolybdate method (Hach method 8185), and low range silica by the heteropoly blue method (Hach method 8186).

Ordering information: Pack of 100 Powder Pillows.